Assay Details
Gene Symbol: LOC101120595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977130.1 | XP_027832931.1 | 33 - 34 | 4384 | 68 | |
XM_027977131.1 | XP_027832932.1 | 34 - 35 | 4294 | 68 | ||
XM_027977133.1 | XP_027832934.1 | 33 - 34 | 4384 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
