Assay Details
Gene Symbol: SCIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185560.3 | XP_012040950.1 | 3 - 4 | 608 | 91 | |
XM_012185561.3 | XP_012040951.1 | 3 - 4 | 476 | 91 | ||
XM_027975082.1 | XP_027830883.1 | 3 - 4 | 610 | 91 | ||
XM_027975083.1 | XP_027830884.1 | 3 - 4 | 591 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
