Assay Details
Gene Symbol: LOC101122940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175443.3 | XP_012030833.2 | 6 - 7 | 728 | 85 | |
XM_012175444.3 | XP_012030834.2 | 6 - 7 | 948 | 85 | ||
XM_012175446.3 | XP_012030836.2 | 6 - 7 | 947 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
