Assay Details
Gene Symbol: CCDC187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966189.1 | XP_027821990.1 | 12 - 13 | 3129 | 65 | |
XM_027966190.1 | XP_027821991.1 | 11 - 12 | 2994 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
