Assay Details
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012697.3 | XP_004012746.1 | 4 - 5 | 637 | 66 | |
XM_027974890.1 | XP_027830691.1 | 4 - 5 | 640 | 66 | ||
XM_027974891.1 | XP_027830692.1 | 4 - 5 | 640 | 66 | ||
XM_027974892.1 | XP_027830693.1 | 4 - 5 | 640 | 66 | ||
XM_027974893.1 | XP_027830694.1 | 4 - 5 | 642 | 66 | ||
XM_027974894.1 | XP_027830695.1 | 4 - 5 | 638 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
