Assay Details
Gene Symbol: MYCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973699.1 | XP_027829500.1 | 23 - 24 | 3665 | 76 |
Gene Symbol: LOC114116662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973851.1 | XP_027829652.1 | 5 - 6 | 541 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
