Assay Details
Gene Symbol: NES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012183418.3 | XP_012038808.2 | 2 - 3 | 1055 | 75 | |
XM_012183429.3 | XP_012038819.2 | 2 - 3 | 1055 | 75 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012183418.3 | XP_012038808.2 | 2 - 3 | 1055 | 75 | |
XM_012183429.3 | XP_012038819.2 | 2 - 3 | 1055 | 75 |