Assay Details
Gene Symbol: APOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015314.4 | XP_004015363.2 | 2 - 3 | 343 | 107 | |
XM_027978462.1 | XP_027834263.1 | 3 - 4 | 2419 | 107 |
Gene Symbol: APOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015313.4 | XP_004015362.2 | 4 - 5 | 1202 | 107 | |
XM_027978463.1 | XP_027834264.1 | 3 - 4 | 2446 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
