Assay Details
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012163196.3 | XP_012018586.1 | 6 - 7 | 673 | 57 | |
XM_015094197.2 | XP_014949683.1 | 6 - 7 | 669 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
