Assay Details
Gene Symbol: LOC101122882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978752.1 | XP_027834553.1 | 4 - 5 | 514 | 61 | |
XM_027978753.1 | XP_027834554.1 | 3 - 4 | 339 | 61 |
Gene Symbol: RDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978806.1 | XP_027834607.1 | 4 - 5 | 514 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
