Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021808.4 | XP_004021857.2 | 4 - 5 | 377 | 63 | |
XM_027962608.1 | XP_027818409.1 | 3 - 4 | 332 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
