Assay Details
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021909.4 | XP_004021958.1 | 19 - 20 | 2799 | 62 | |
XM_012106233.3 | XP_011961623.1 | 19 - 20 | 2804 | 62 | ||
XM_012106235.3 | XP_011961625.1 | 19 - 20 | 2731 | 62 | ||
XM_027963598.1 | XP_027819399.1 | 18 - 19 | 2705 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
