Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967817.1 | XP_027823618.1 | 12 - 13 | 1021 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967817.1 | XP_027823618.1 | 12 - 13 | 1021 | 114 |
Set Membership: |
![]() ![]() ![]() ![]() |