Assay Details
Gene Symbol: GOLT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186925.3 | XP_012042315.2 | 3 - 4 | 406 | 110 | |
XM_012186926.3 | XP_012042316.2 | 3 - 4 | 339 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
