Assay Details
Gene Symbol: LOC101120628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963716.1 | XP_027819517.1 | 1 - 2 | 138 | 104 |
Gene Symbol: LOC114117583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976790.1 | XP_027832591.1 | 1 - 2 | 138 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
