Assay Details
Gene Symbol: RSKR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013227.4 | XP_004013276.1 | 2 - 3 | 602 | 91 | |
XM_012185397.3 | XP_012040787.1 | 2 - 3 | 602 | 91 | ||
XM_015098538.2 | XP_014954024.1 | 2 - 3 | 602 | 91 | ||
XM_015098539.2 | XP_014954025.1 | 2 - 3 | 602 | 91 | ||
XM_027975232.1 | XP_027831033.1 | 2 - 3 | 602 | 91 | ||
XM_027975233.1 | XP_027831034.1 | 2 - 3 | 602 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
