Assay Details
Gene Symbol: TMPRSS11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180072.3 | XP_012035462.1 | 8 - 9 | 1260 | 103 | |
XM_012180073.3 | XP_012035463.1 | 7 - 8 | 1221 | 103 | ||
XM_012180074.3 | XP_012035464.1 | 8 - 9 | 1218 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
