Assay Details
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098902.2 | XP_014954388.2 | 7 | 1565 | 118 | |
XM_027974294.1 | XP_027830095.1 | 7 | 1547 | 118 | ||
XM_027974295.1 | XP_027830096.1 | 7 | 1517 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
