Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976754.1 | XP_027832555.1 | 4 - 5 | 595 | 61 | |
XM_027976757.1 | XP_027832558.1 | 4 - 5 | 595 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
