Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012133548.3 | XP_011988938.1 | 3 - 4 | 547 | 97 | |
XM_027964843.1 | XP_027820644.1 | 3 - 4 | 444 | 97 | ||
XM_027964844.1 | XP_027820645.1 | 3 - 4 | 329 | 97 | ||
XM_027964845.1 | XP_027820646.1 | 3 - 4 | 401 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
