Assay Details
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011339.4 | XP_004011388.3 | 5 - 6 | 1215 | 73 | |
GenBank mRNA | KC425611.1 | - | 5 - 6 | 1221 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
