Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009036.3 | XP_004009085.2 | 5 - 6 | 614 | 97 | |
XM_012178886.3 | XP_012034276.2 | 5 - 6 | 589 | 97 | ||
XM_012178887.2 | XP_012034277.2 | 5 - 6 | 611 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
