Assay Details
Gene Symbol: SELENOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979805.1 | XP_027835606.1 | 2 - 3 | 431 | 96 | |
XM_027979806.1 | XP_027835607.1 | 2 - 3 | 439 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
