Assay Details
Gene Symbol: CPSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961170.1 | XP_027816971.1 | 1 - 2 | 262 | 67 | |
XM_027961171.1 | XP_027816972.1 | 1 - 2 | 274 | 67 | ||
XM_027961172.1 | XP_027816973.1 | 1 - 2 | 281 | 67 | ||
XM_027961173.1 | XP_027816974.1 | 1 - 2 | 262 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
