Assay Details
Gene Symbol: CARNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964491.1 | XP_027820292.1 | 6 - 7 | 1910 | 68 | |
XM_027964493.1 | XP_027820294.1 | 6 - 7 | 2182 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
