Assay Details
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004081.3 | XP_004004130.1 | 5 - 6 | 752 | 78 | |
XM_004004082.4 | XP_004004131.1 | 5 - 6 | 784 | 78 | ||
XM_012116286.3 | XP_011971676.1 | 5 - 6 | 1409 | 78 | ||
XM_012116293.2 | XP_011971683.1 | 5 - 6 | 746 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
