Assay Details
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960505.1 | XP_027816306.1 | 4 - 5 | 619 | 59 | |
XM_027960507.1 | XP_027816308.1 | 4 - 5 | 586 | 59 | ||
XM_027960508.1 | XP_027816309.1 | 4 - 5 | 586 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
