Assay Details
Gene Symbol: LOC101106395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004155.4 | XP_004004204.1 | 4 - 5 | 1054 | 66 | |
XM_027964301.1 | XP_027820102.1 | 4 - 5 | 1054 | 66 | ||
XM_027964302.1 | XP_027820103.1 | 4 - 5 | 1054 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
