Assay Details
Gene Symbol: LOC101106395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004155.4 | XP_004004204.1 | 7 - 8 | 1432 | 78 | |
XM_027964301.1 | XP_027820102.1 | 8 - 9 | 1558 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
