Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021523.4 | XP_004021572.1 | 3 - 4 | 446 | 99 | |
XM_004021524.3 | XP_004021573.1 | 3 - 4 | 298 | 99 | ||
XM_004021525.4 | XP_004021574.1 | 3 - 4 | 417 | 99 | ||
XM_012105420.2 | XP_011960810.1 | 3 - 4 | 280 | 99 | ||
XM_012105423.3 | XP_011960813.1 | 3 - 4 | 827 | 99 | ||
XM_027962416.1 | XP_027818217.1 | 3 - 4 | 293 | 99 | ||
XM_027962418.1 | XP_027818219.1 | 3 - 4 | 509 | 99 | ||
XM_027962419.1 | XP_027818220.1 | 3 - 4 | 480 | 99 | ||
XM_027962420.1 | XP_027818221.1 | 4 - 5 | 630 | 99 | ||
XM_027962421.1 | XP_027818222.1 | 3 - 4 | 421 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
