Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021782.4 | XP_004021831.1 | 3 - 4 | 1293 | 64 | |
XM_012105930.3 | XP_011961320.1 | 3 - 4 | 1293 | 64 | ||
XM_012105933.3 | XP_011961323.1 | 3 - 4 | 1293 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
