Assay Details
Gene Symbol: TOGARAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012099164.3 | XP_011954554.3 | 3 - 4 | 2532 | 105 | |
XM_027957299.1 | XP_027813100.1 | 3 - 4 | 2532 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
