Assay Details
Gene Symbol: TASOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099687.2 | XP_014955173.2 | 19 - 20 | 11478 | 72 | |
XM_015099688.2 | XP_014955174.2 | 19 - 20 | 11459 | 72 | ||
XM_015099691.2 | XP_014955177.2 | 18 - 19 | 11234 | 72 | ||
XM_027976780.1 | XP_027832581.1 | 19 - 20 | 11478 | 72 | ||
XM_027976781.1 | XP_027832582.1 | 20 - 21 | 7908 | 72 | ||
XM_027976782.1 | XP_027832583.1 | 13 - 14 | 6601 | 72 | ||
XM_027976783.1 | XP_027832584.1 | 18 - 19 | 11216 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
