Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 20 - 21 | 2907 | 92 | |
XM_027975770.1 | XP_027831571.1 | 20 - 21 | 2906 | 92 | ||
XM_027975771.1 | XP_027831572.1 | 20 - 21 | 2878 | 92 | ||
XM_027975772.1 | XP_027831573.1 | 19 - 20 | 2710 | 92 | ||
XM_027975773.1 | XP_027831574.1 | 11 - 12 | 1357 | 92 | ||
XM_027975774.1 | XP_027831575.1 | 11 - 12 | 1236 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
