Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 21 - 22 | 2992 | 69 | |
XM_027975770.1 | XP_027831571.1 | 21 - 22 | 2991 | 69 | ||
XM_027975771.1 | XP_027831572.1 | 21 - 22 | 2963 | 69 | ||
XM_027975772.1 | XP_027831573.1 | 20 - 21 | 2795 | 69 | ||
XM_027975773.1 | XP_027831574.1 | 12 - 13 | 1442 | 69 | ||
XM_027975774.1 | XP_027831575.1 | 12 - 13 | 1321 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
