Assay Details
Gene Symbol: LOC101104843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975767.1 | XP_027831568.1 | 22 - 23 | 3114 | 70 | |
XM_027975770.1 | XP_027831571.1 | 22 - 23 | 3113 | 70 | ||
XM_027975771.1 | XP_027831572.1 | 22 - 23 | 3085 | 70 | ||
XM_027975772.1 | XP_027831573.1 | 21 - 22 | 2917 | 70 | ||
XM_027975773.1 | XP_027831574.1 | 13 - 14 | 1564 | 70 | ||
XM_027975774.1 | XP_027831575.1 | 13 - 14 | 1443 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
