Assay Details
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005829.4 | XP_004005878.2 | 5 - 6 | 2696 | 96 | |
XM_027966687.1 | XP_027822488.1 | 6 - 7 | 2602 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
