Assay Details
Gene Symbol: LOC101103343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015280.4 | XP_004015329.1 | 5 - 6 | 1248 | 127 | |
XM_027978376.1 | XP_027834177.1 | 5 - 6 | 1327 | 127 | ||
XM_027978377.1 | XP_027834178.1 | 5 - 6 | 1137 | 127 | ||
XM_027978378.1 | XP_027834179.1 | 5 - 6 | 1321 | 127 | ||
XM_027978379.1 | XP_027834180.1 | 5 - 6 | 1321 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
