Assay Details
Gene Symbol: LOC101106975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957845.1 | XP_027813646.1 | 7 - 8 | 700 | 148 | |
XM_027957846.1 | XP_027813647.1 | 9 - 10 | 815 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() |
