Assay Details
Gene Symbol: LOC114109610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958637.1 | XP_027814438.1 | 2 - 3 | 222 | 79 |
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 14 - 15 | 1962 | 79 | |
XM_012101169.3 | XP_011956559.1 | 13 - 14 | 1924 | 79 | ||
XM_027958396.1 | XP_027814197.1 | 12 - 13 | 2022 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
