Assay Details
Gene Symbol: LOC114109610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958637.1 | XP_027814438.1 | 3 - 4 | 346 | 112 |
Gene Symbol: LOC101106806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012101164.3 | XP_011956554.1 | 15 - 16 | 2086 | 112 | |
XM_012101169.3 | XP_011956559.1 | 14 - 15 | 2048 | 112 | ||
XM_027958396.1 | XP_027814197.1 | 13 - 14 | 2146 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
