Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975168.1 | XP_027830969.1 | 21 - 22 | 2366 | 75 | |
XM_027975170.1 | XP_027830971.1 | 20 - 21 | 2285 | 75 | ||
XM_027975171.1 | XP_027830972.1 | 21 - 22 | 2452 | 75 | ||
XM_027975172.1 | XP_027830973.1 | 6 - 7 | 744 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
