Assay Details
Gene Symbol: LOC101113775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974198.1 | XP_027829999.1 | 6 - 7 | 984 | 98 | |
XM_027974199.1 | XP_027830000.1 | 5 - 6 | 790 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
