Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976299.1 | XP_027832100.1 | 9 - 10 | 2241 | 115 | |
XM_027976301.1 | XP_027832102.1 | 10 - 11 | 1908 | 115 | ||
XM_027976302.1 | XP_027832103.1 | 9 - 10 | 1896 | 115 | ||
XM_027976303.1 | XP_027832104.1 | 9 - 10 | 1945 | 115 | ||
XM_027976304.1 | XP_027832105.1 | 7 - 8 | 1777 | 115 | ||
XM_027976305.1 | XP_027832106.1 | 9 - 10 | 1545 | 115 | ||
XM_027976307.1 | XP_027832108.1 | 8 - 9 | 1434 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
