Assay Details
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012188692.3 | XP_012044082.2 | 15 - 16 | 2299 | 153 |
Gene Symbol: LOC114112487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963804.1 | XP_027819605.1 | 1 - 2 | 123 | 153 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027963804.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
