Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013657.4 | XP_004013706.1 | 1 - 2 | 224 | 89 | |
XM_012187191.3 | XP_012042581.1 | 1 - 2 | 224 | 89 | ||
XM_012187193.3 | XP_012042583.1 | 1 - 2 | 224 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
