Assay Details
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097907.3 | XP_011953297.1 | 4 - 5 | 485 | 111 | |
XM_012097915.3 | XP_011953305.1 | 4 - 5 | 485 | 111 | ||
XM_027957637.1 | XP_027813438.1 | 4 - 5 | 485 | 111 | ||
XM_027957641.1 | XP_027813442.1 | 3 - 4 | 384 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027957641.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
