Assay Details
Gene Symbol: MIR376B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107908.1 | 1 | 44 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-654 | MIR654 | oar-mir-654 | -122.0 | Sense | Chr. 18 - 66455659 - 66455763 on NCBI Build Oar_rambouillet_v1.0 |
Downstream | microRNA mir-376a | MIR376A | oar-mir-376a | 274.0 | Sense | Chr. 18 - 66456264 - 66456366 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
