Assay Details
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272334.2 | XP_008270556.1 | 1 - 2 | 2646 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272334.2 | XP_008270556.1 | 1 - 2 | 2646 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |