Assay Details
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082311.1 | NP_001075780.1 | 3 - 4 | 459 | 101 | |
GenBank mRNA | M86381.1 | - | 3 - 4 | 459 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
